Negative Carriers, Electron Microscopy Sciences
Product Details & Documents
Universal glass-type masking negative carriers features four individually adjustable masking blades and removable glass inserts
Glassless, sandwich type negative carriers are rofessional quality, with black, anodized aluminum construction. Hinged for quick handling.
Universal glass-type masking negative carriers features four individually adjustable masking blades and removable glass inserts. Accepts accessory anti-Newton glass. Top and bottom glass can be removed to convert it to a single-size glassless carrier by means of lock-in inserts.
Accepts accessory anti-Newton glass. Top and bottom glass can be removed to convert it to a single-size glassless carrier by means of lock-in inserts.
Specifications
- Description:Universal Glass-Type