Orders LinkContactUsLinkComponent
Light Boxes:, Electron Microscopy Sciences
Light Boxes:, Electron Microscopy Sciences
Replacement Light Bulb for Model B
Catalog # 100493-422
Light Boxes:, Electron Microscopy Sciences
Catalog # 100493-422
Supplier:  
Light Boxes:, Electron Microscopy Sciences
Light Boxes:, Electron Microscopy Sciences
Replacement Light Bulb for Model B
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Model A: It is compact and can be used on a desk
For viewing and editing slides, transparencies, negatives, micrographs, etc. They also serve as a base for orientation of embedding specimens when observing tissues using a magnifier. Scientifically designed to provide even distribution and uniformity of light. All metal welded construction with a white baked-enamel finish. The viewing surface is a translucent acrylic that is break resistant. It can be used flat on a table, inclined, standing, or hung on the wall. It comes with a ANSI standard 5000K fluorescent lamp, 10 feet cord, on-off switch.
Model A: It is compact and can be used on a desk. Small enough to carry in a briefcase or store in a drawer. Weight 4.5 lbs. Viewing surface measures: 22 x 29.2 cm (8 3/4 x 11 1/2"). Overall measurements: 24.1 x 31.6 x 5 cm (9 1/2 x 12 1/2 x 2").
Small enough to carry in a briefcase or store in a drawer. Weight 4.5 lbs. Viewing surface measures: 22 x 29.2 cm (8 3/4 x 11 1/2"). Overall measurements: 24.1 x 31.6 x 5 cm (9 1/2 x 12 1/2 x 2").
Specifications
  • Description:
    Replacement Light Bulb for Model B