Orders LinkContactUsLinkComponent
Pyrex® Plate, Electron Microscopy Sciences
Pyrex® Plate, Electron Microscopy Sciences
Pyrex Plate
Catalog # 100493-410
Supplier #:  71563-01
Pyrex® Plate, Electron Microscopy Sciences
Catalog # 100493-410
Pyrex® Plate, Electron Microscopy Sciences
Pyrex® Plate, Electron Microscopy Sciences
Pyrex Plate
Price
$147.79
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A 9 cavity Pyrex pressed plate which offers a clear view for observation by transmitted light.
The plate measures L×W: 100×85 mm (4×3⅜"). The cavity is 6.4 mm (¼") deep with a 22 mm (⅞") opening.
Specifications
  • Description:
    Pyrex Plate