Orders LinkContactUsLinkComponent
Capsule Molds, Electron Microscopy Sciences
Capsule Molds, Electron Microscopy Sciences
Capsule Mold Type C, 12 Cavities
Catalog # 100491-600
Supplier #:  70161
Capsule Molds, Electron Microscopy Sciences
Catalog # 100491-600
Capsule Molds, Electron Microscopy Sciences
Capsule Molds, Electron Microscopy Sciences
Capsule Mold Type C, 12 Cavities
Price
$44.66
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Our molds are made from the strongest possible silicone rubber providing extremely high tear resistance. They are all reusable and produce embedding blocks of numerous shapes and sizes with a wide variety of tips. We also offer a mold which produces blocks to be used for re-embedding.
Specifications
  • Description:
    Capsule Mold Type C, 12 Cavities