Orders LinkContactUsLinkComponent
Polystyrene Containers, Electron Microscopy Sciences
Polystyrene Containers, Electron Microscopy Sciences
30 ml, Container with Spoon, 93.1 mm, PP, universal cap, Plain, 30.1 mm
Catalog # 100490-480
Polystyrene Containers, Electron Microscopy Sciences
Catalog # 100490-480
Supplier:  
Polystyrene Containers, Electron Microscopy Sciences
Polystyrene Containers, Electron Microscopy Sciences
30 ml, Container with Spoon, 93.1 mm, PP, universal cap, Plain, 30.1 mm
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These clear and rigid containers are made from polystyrene.
  • All containers are aseptically manufactured
  • Labeling options include unlabeled, printed, or plain
  • 30mL containers with universal closure rely on special seal technology
  • Other sizes employ liners or crab claw closures to facilitate sample containment
  • Some sizes are available with spoons
  • For in vitro use only
Specifications
  • Cap type:
    PP, universal cap
  • Ø:
    30.1 mm
  • Capacity:
    30 ml
  • Description:
    Container with Spoon
  • Height:
    93.1 mm
  • Label type:
    Plain