Orders LinkContactUsLinkComponent
Dropping Bottles, Glass, Electron Microscopy Sciences
Dropping Bottles, Glass, Electron Microscopy Sciences
Clear Dropping Bottle, 50 ml
Catalog # 100490-426
Supplier #:  64220-05
Dropping Bottles, Glass, Electron Microscopy Sciences
Catalog # 100490-426
Dropping Bottles, Glass, Electron Microscopy Sciences
Clear Dropping Bottle, 50 ml
Price
$160.05
Log in for availability
Pack of 4
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These bottles deliver stains and indicator solutions drop by drop from a projecting drip tip on the side of the stopper.
A partial turn of the ground glass stopper will seal the bottle.
Specifications
  • Description:
    Clear Dropping Bottle, 50 ml