Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Gold Sol, Electron Microscopy Sciences

Gold Sol, Electron Microscopy Sciences

Gold Sol, 100 ml, 15 nm
Catalog # 100487-930
Minority or Woman-Owned Business Enterprise
Supplier #:  25516
Gold Sol, Electron Microscopy Sciences
Catalog # 100487-930
Minority or Woman-Owned Business Enterprise
CAS Number:  
Gold Sol, Electron Microscopy Sciences
Gold Sol, Electron Microscopy Sciences

Gold Sol, 100 ml, 15 nm

CAS Number:  
Price
$443.48
Log in for availability
Each (100ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Solutions of high quality unconjugated gold particles.

  • Particle sizes of 0.8, 6, 10 ,15 and 25 nm
  • Guaranteed shelf life of 12 months
  • Monodisperse particle population

To prepare a high quality (immuno) gold conjugate it is important to have particles with uniform size and highly adsorptive surfaces.

Introduction
The preparation of conventional gold reagents is based on gold particles with a diameter suited for direct electron microscopic visualization. Aurion offers a range of these Reagents which cover most of the approaches in transmission and scanning electron microscopy.
Aurion Gold Sols (solutions of high quality unconjugated gold particles) provide opportunities for users to prepare conjugates with primary antibodies, ligands and other binding agents with the same particle characteristics as in the Conventional Immunogold Reagents.

Product Description
AURION Gold Sols are prepared according to a unique protocol, warranting narrow size distribution and reproducible adsorption characteristics.
The AURION gold sols are available in the same size range as the Conventional Immunogold Reagents: 6, 10, 15 and 25 nm. The particle population is monodispersed and thus shows minimal size variation and overlap. Typically, the coefficient of variance for the 6 and 25 nm particle size sols is less than 12%, whereas the 10 and 15 nm size sols show less than 10% variation. Actual lot specifications (size, variation and expiry date) are indicated on the accompanying package insert.
Package size: 100 ml of high quality gold sol at an OD520 nm of approximately 1.

Storage
AURION gold sols have a guaranteed shelf life of 12 months from the date of quality control analysis.
The products should be stored at 4 – 8°C.
Freezing is not recommended.

Specifications

Specifications

  • Description:
    Gold Sol
  • Size:
    15 nm
  • Volume:
    100 ml
  • Cat. No.:
    100487-930
  • Antibody Type:
    Secondary

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.