Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Iron Standards

10000 µg/ml, 125 ml, Iron, HNO₃ dil., Clear/Light Tan
Catalog # 100359-992
Marketsource
 :  CGFE10-125ML
Iron Standards
Catalog # 100359-992
Supplier:  
Marketsource
CAS Number:  
Iron Standards
Iron Standards

10000 µg/ml, 125 ml, Iron, HNO₃ dil., Clear/Light Tan

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Certified reference material for Iron, traceable to NIST.

One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).

 : ISO 17025/17034.

Specifications

Specifications

  • Concentration:
    10000 µg/ml
  • Matrix:
    HNO₃ dil.
  • Color:
    Clear/Light Tan
  • Size:
    125 ml
  • Element:
    Iron

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.