Open Case Carts, Lakeside®
Sold shelf with divider for use with open case cart
Catalog # 89210-182
Supplier: Lakeside
: 7940SD
Jump to:
Product Details & Documents
Open case carts can be configured to meet your every need
Maximum design flexibility for your best fit. Multiple uses as case carts, supply carts, distribution and linen carts. Stainless steel frame is easy to clean and sanitize. Solid stainless steel or wire shelves allow options for different size supplies. All-welded construction eliminates nuts and bolts for a stronger structure and easier to clean surface. Fits elevators and adaptable for cart pick-up systems. 6" casters for easy maneuverability. Easy to configure with 51/2" space between shelf rungs. No tools required for assembly.
Specifications
- Description:Sold shelf with divider for use with open case cart