Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VWR® Basic SPP Coveralls, Blue

VWR® Basic SPP Coveralls, Blue

Blue, L, 38¾" (body), 29½" (leg), 25"
Catalog # 89208-738
Supplier:   VWR International
VWR® Basic SPP Coveralls, Blue
Catalog # 89208-738
Supplier:  VWR International
VWR® Basic SPP Coveralls, Blue
VWR® Basic SPP Coveralls, Blue

Blue, L, 38¾" (body), 29½" (leg), 25"

Price
$84.43
Log in for availability
Case of 25
Product Details & Documents

Disposable coverall made from spunbond polypropylene (SPP) fabric, offering lightweight and breathable protection for non-hazardous environments. Ideal for general industrial use, food processing, and clean areas requiring basic contamination control.

  • Lightweight and breathable fabric minimizes heat buildup

  • Zipper front closure for easy donning and removal
  • Elastic wrists and ankles for secure coverage
  • Disposable, single-use design for contamination control
  • Economical solution for basic protection needs
  • Non-linting material helps maintain cleanliness
  • Ideal for general industrial, food processing, and nonhazardous environments

The Basic Protection SPP Coverall is an economical and practical solution for environments requiring lightweight, disposable protective apparel. Constructed from spunbond polypropylene (SPP) fabric, this garment offers a balance of comfort and functionality. The non-woven material is breathable and non-linting, making it suitable for clean areas, food processing, and general industrial applications where contamination control is essential but exposure risks are minimal.

Designed for ease of use, the coverall features a zipper front closure for quick donning and removal, along with elastic wrists and ankles to ensure a secure fit and reduce gaps. The single-use design supports hygiene compliance and simplifies inventory management, while the lightweight construction minimizes heat buildup during extended wear.

Although not intended for hazardous chemical or liquid protection, the Basic Protection SPP Coverall provides an effective barrier against dust, dirt, and dry particulates. This makes it an ideal choice for controlled environments, pre-gowning areas, and routine operations where basic protection is required. By combining cost-efficiency, comfort, and contamination control, this coverall delivers dependable performance for facilities seeking reliable disposable apparel for everyday tasks.

Packaging: Coveralls are packaged 25 per case.

Specifications
  • Clothing Size:
    L
  • Color:
    Blue
  • Length:
    38¾" (body), 29½" (leg)
  • Waist size:
    25"
  • Tensile Strength:
    16.1 (Filling)
  • Air Permeability:
    95.3
  • Bacterial Filtration Efficiency:
    0.9
  • Sleeve or leg length:
    Long
  • Tearing Strength:
    2.6 (Widthwise Yarns)
  • Gender:
    Unisex
  • Closure type:
    With Zip Closure
  • Weight:
    30 g/m²
  • H₂O Resistance:
    16.7
  • Cat. No.:
    89208-738
  • Water Vapor Transmission Rate:
    965
  • Cleanroom Class:
    10000/ISO 7
  • Garment application:
    Cleanroom|Industrial|Laboratory
  • Pockets:
    Without Pockets
  • Fabric:
    Spunbound polypropylene
  • Particle shedding (Helmke Drum):
    Level II
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.