Order Entry
Puerto Rico
ContactUsLinkComponent
HPLC COL 100 X 3.2MM 5U 100A
Catalog # 95005-204
Supplier:  PerkinElmer
undefined
HPLC COL 100 X 3.2MM 5U 100A
Catalog # 95005-204
Supplier:  PerkinElmer
Supplier Number:  12D211D

Related Information