Orders LinkContactUsLinkComponent
AlphaMop™ Polyester Mop
AlphaMop™ Polyester Mop
Pre-wetted covers and pads, 10 covers and 1 pad/bag; 12 bags/case
Catalog # TW-TX7026
Supplier:   Texwipe
Supplier #:  TX7026
AlphaMop™ Polyester Mop
Catalog # TW-TX7026
Supplier:  Texwipe
AlphaMop™ Polyester Mop
AlphaMop™ Polyester Mop
Pre-wetted covers and pads, 10 covers and 1 pad/bag; 12 bags/case
Price
$243.00
Log in for availability
Pack of 25 Case of 100
Product Details & Documents
Ideal for cleaning walls, floors, and other flat surfaces in critical environments.
AlphaMop™ covers TWTX7026 are pre-wetted with 6% isopropyl alcohol. The case purchase includes two foam pads. Replacement pack TW-TX7118 contains 150 polyester heads and 6 foam pads.
The changeable, laundered polyester head fits over a flat foam swivel pad assembly attached to a fiberglass handle. Replaceable foam pad ensures the polyester head conforms to textured floors and other surfaces while resisting corrosion and abrasion. Lightweight mop maintains ultralow generation of particles and extractables. Bag-within-a-Bag® packaged.
Specifications
  • Description:
    Pre-wetted covers and pads
  • Packed:
    10 covers and 1 pad/bag; 12 bags/case