Congo red 0,1% (w/v) in aqueous solution indicator
Poly bottle, 120 mL
Supplier: Ricca Chemical
Supplier #: R2250000-120A
Congo red 0,1% (w/v) in aqueous solution indicator
Catalog # RC22504
Supplier: Ricca Chemical
CAS Number: 573-58-0
Jump to:
Product Details & Documents
Synonyms: CI 22120
- CAS Number:573-58-0
- Formula:C₃₂H₂₂N₆Na₂O₆S₂
- MDL Number:MFCD00004028
- Molecular Weight:696.67 g/mol
- Boiling Pt:100 °C
- Melting Pt:0.0 °C
- Density:1.0010 g/cm³
- Storage Temperature:Ambient
Documents
Specifications
Specification Test Results
- Appearance:Clear, red, odorless
- Suitability for Use:Red (pH > 5.0) - Blue (pH < 3.0)
Specifications
- Pack type:Poly bottle
- Size:120 mL
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



