KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences
MICROFLEX® threaded erlenmeyer flasks, 25 ml, 60 mm, 20 - 400
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 747530-2520
KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences
Catalog # KT747530-2520
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
These threaded Erlenmeyer flasks are useful for sample collection, derivatization reactions, extractions, sample storage and micro mixing.
- Without graduations
- Manufactured from 33 expansion, low extractable borosilicate glass
Accessory for Microscale MICROFLEX™ threaded kits.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Documents
Certificates
Search by Lot NumberSpecifications
- Capacity:25 ml
- Description:MICROFLEX® threaded erlenmeyer flasks
- Height:60 mm
- Thread:20 - 400
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



