Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences

KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences

MICROFLEX® threaded erlenmeyer flasks, 10 ml, 45 mm, 20 - 400
Catalog # KT747530-1020
Supplier #:  747530-1020
KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences
Catalog # KT747530-1020
KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences
KIMBLE® MICROFLEX® Erlenmeyer Flasks, Threaded, DWK Life Sciences

MICROFLEX® threaded erlenmeyer flasks, 10 ml, 45 mm, 20 - 400

Price
$34.10
Log in for availability
Each
Product Details & Documents

These threaded Erlenmeyer flasks are useful for sample collection, derivatization reactions, extractions, sample storage and micro mixing.

  • Without graduations
  • Manufactured from 33 expansion, low extractable borosilicate glass

Accessory for Microscale MICROFLEX™ threaded kits.

 : Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Documents

Specifications
  • Capacity:
    10 ml
  • Description:
    MICROFLEX® threaded erlenmeyer flasks
  • Height:
    45 mm
  • Thread:
    20 - 400
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.