Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Microcentrifuge Tubes, Sorenson BioScience

Microcentrifuge Tubes, Sorenson BioScience

Microcentrifuge tube 1.7 ml, low binding, 1.7 ml, Natural, Non sterile
Catalog # 89082-332
Supplier:   Sorenson Bioscience
Supplier #:  39640T
Microcentrifuge Tubes, Sorenson BioScience
Catalog # 89082-332
Microcentrifuge Tubes, Sorenson BioScience
Microcentrifuge Tubes, Sorenson BioScience

Microcentrifuge tube 1.7 ml, low binding, 1.7 ml, Natural, Non sterile

Price
$35.88
Log in for availability
Pack of 250 Case of 2500
Product Details & Documents

SafeSeal™ Microcentrifuge tubes are ideal for high-speed centrifugation, rated up to 25000×g.

  • Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors

A positive seal design allows repeated cap closures and will not pop during boiling or freezing. Tubes feature external graduation marks and frosted writing surfaces; fit all standard rotors; are autoclavable; and are rated from −80 to 121 °C.

Specifications
  • Sterility:
    Non sterile
  • Volume:
    1.7 ml
  • Color:
    Natural
  • Description:
    Microcentrifuge tube 1.7 ml, low binding
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.