Adapters, Distilling, Process Reactors, Angled, Chemglass
Distilling Adapter, Double, 10° Angled Side Neck, 1/2" Swagelok, 24/40, 6 mm (PTFE)
Catalog # 89028-406
Supplier: Chemglass
Supplier #: CG-1235-80
Jump to:
Product Details & Documents
Adapter is designed for use with flasks or reaction vessel lids with a 10 or 20° angled side neck.
- Adapter can be used with compound angled side necks
- Five neck vessel lids and vessel lid sets are designed for use with this adapter
Adapter has a lower inner joint and two top outer joints. Lower stopcock is either a 6 mm or a 10 mm PTFE stopcock with a 1/2" or 3/4" O.D. distillate takeoff for use with a compression fitting and TFE tubing.
Specifications
- Description:Distilling Adapter, Double, 10° Angled Side Neck, 1/2" Swagelok
- Stopcock size:6 mm (PTFE)
- Joint size:24/40
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



