Orders LinkContactUsLinkComponent
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
250 ml, 24/40, 83 x 140 mm
Catalog # 89001-286
Supplier #:  25285-250
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
Catalog # 89001-286
Supplier:  
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
250 ml, 24/40, 83 x 140 mm
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FLASK BOIL RND BOT 250 ML PK2 Catalog #89090-798
Product 89090-798 is similar to 89001-286 but manufacturer and specifications may vary.
FLASK BOIL VWR RND BTM BORO 3.3 GL 250ML Catalog #10536-666
Product 10536-666 is similar to 89001-286 but manufacturer and specifications may vary.
FLASK HW RB 24/40 250ML Catalog #80068-778
Product 80068-778 is similar to 89001-286 but manufacturer and specifications may vary.
See more alternatives
Product Details & Documents
Short neck flasks are designed to ASTM Specification E1403, Class 3 requirements
They feature a full-length, outer, [ST] 24/40 joint.
Documents
Specifications
  • [ST] joint no.:
    24/40
  • Capacity:
    250 ml
  • O.D.×H:
    83 x 140 mm