Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VITLAB® Graduated Pitchers, SAN, BrandTech

VITLAB® Graduated Pitchers, SAN, BrandTech

500 ml (17 oz.), 10 ml
Catalog # 83008-738
Supplier:   Brandtech
Supplier #:  44191
VITLAB® Graduated Pitchers, SAN, BrandTech
Catalog # 83008-738
Supplier:  Brandtech
VITLAB® Graduated Pitchers, SAN, BrandTech
VITLAB® Graduated Pitchers, SAN, BrandTech

500 ml (17 oz.), 10 ml

Price
$198.74
Log in for availability
Pack of 12
Minimum order quantity is 1
Product Details & Documents

Clear SAN (styrene acrylonitrile copolymer) pitchers with molded graduations are a lightweight, safer alternative to glass in many applications.

  • Glass-clear pitchers have contoured handles for easy lifting
  • Durable molded graduations for long measuring life
  • Pouring lip reduces drips
  • SAN has a maximum temperature for use of 70 ˚C

Enhanced graduations are provided at major intervals for easy volume estimations.

Specifications
  • Graduation Interval:
    10 ml
  • Capacity:
    500 ml (17 oz.)