Orders LinkContactUsLinkComponent
Bushing Adapters, Chemglass
Bushing Adapters, Chemglass
Bushing Adapter, 14/20, 10/18
Catalog # 80065-788
Supplier:   Chemglass
Supplier #:  CG-1016-27
Bushing Adapters, Chemglass
Catalog # 80065-788
Supplier:  Chemglass
Bushing Adapters, Chemglass
Bushing Adapters, Chemglass
Bushing Adapter, 14/20, 10/18
Price
$65.73
Log in for availability
Each
Product Details & Documents
Bushing style adapters feature a lower standard taper inner joint and outer joint ground in the bore of the adapter.
  • Used to insert thermometers into apparatus
  • Joint sizes as listed
Adapters have a 10/18 or 10/30 outer member in the bore.
Specifications
  • Description:
    Bushing Adapter
  • [ST] inner joint:
    14/20
  • [ST] outer joint:
    10/18