Orders LinkContactUsLinkComponent
Trypsinizing Flasks, with Cap, Wheaton
Trypsinizing Flasks, with Cap, Wheaton
50 ml, 24-430
Catalog # 62401-624
Supplier #:  355392
Trypsinizing Flasks, with Cap, Wheaton
Catalog # 62401-624
Trypsinizing Flasks, with Cap, Wheaton
Trypsinizing Flasks, with Cap, Wheaton
50 ml, 24-430
Price
$78.10
Log in for availability
Case of 1
Minimum order quantity is 1
Product Details & Documents
Wheaton-33® low extractable borosilicate glass composition with narrow lip
Deep uniform baffles provide excellent turbulence. Internal dam reduces spill out of untrypsinized particles. Black screw cap with white (styrene-butadiene) rubber liner.
Specifications
  • Cap Size:
    24-430
  • Capacity:
    50 ml