Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

VWR® Plug-Type Caps, PE

White
  60819-014
   Sustainable Packaging|Low Manufacturing Impact
VWR® Plug-Type Caps, PE
  60819-014
   Sustainable Packaging|Low Manufacturing Impact
VWR® Plug-Type Caps, PE
VWR® Plug-Type Caps, PE

White

 
$190.11
   
Pack of 1,000 Case of 3000
 

Disposable caps are for 12 x 75 mm and 13 x 100 mm culture tubes.

  • Disposable
  • Polyethylene construction
  • Unique fluted design
  • Snugly grasps plastic or glass tubes

They feature a unique fluted design that grips plastic or glass tubes snugly.

 : The assorted pack contains 500 caps each in yellow, orange, red, blue, green and purple.

 
  • Color:
    White
  • Cat. No.:
    60819-014
  • Environmentally Preferable:
    Sustainable Packaging|Low Manufacturing Impact