Stopcocks, plain bore, PYREX® Rotaflo®
8×215 mm, 0 - 3 mm (0 - 0.118")
Supplier: Corning
: 7475-3
Jump to:
Product Details & Documents
PYREX®, borosilicate glass.
- Fine flow adjustment
- Lubrication is not required
- Fast assembly
These high performance Rotaflo® stopcocks can be adjusted with bore openings and have sidearms. Only chemically resistant PTFE and PYREX® borosilicate glass are in contact with liquids and gase. Seal is visible when open or closed.
Certificates
Search by Lot NumberSpecifications
- Sidearm Dimensions (Ø×L):8×215 mm
- Bore size:0 - 3 mm (0 - 0.118")



