Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
WIPER MS 1200 9X9 PRESAT 7030
Catalog # 47727-520
Supplier:
Berkshire
SDS
Expand image
WIPER MS 1200 9X9 PRESAT 7030
Catalog # 47727-520
Supplier:
Berkshire
Supplier Number:
SPXM12000112
SDS
Related Information
Documentation
SDS
ghs_SPX100000112
Documentation