Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VWR® Powered Backsplash

VWR® Powered Backsplash

Powered backsplash, 15 A, 72"
Catalog # 37000-634
Supplier:   Kewaunee
Supplier #:  KL786150LN
VWR® Powered Backsplash
Catalog # 37000-634
Supplier:  Kewaunee
VWR® Powered Backsplash

Powered backsplash, 15 A, 72"

Price
$1,324.59
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

15 A electrical plugs for 48 or 72" benches.

  • 15 A
  • Backsplash

Power receptacles integrated into the backsplash.

Specifications
  • Description:
    Powered backsplash
  • Length:
    72"
  • Electrical:
    15 A
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.