Orders LinkContactUsLinkComponent
Evolution® High-Column Workstations, Kewaunee®
Evolution® High-Column Workstations, Kewaunee®
Wallaby plastic laminate work surface, 36", 72×30"
Catalog # VC3048WA
Evolution® High-Column Workstations, Kewaunee®
Catalog # VC3048WA
Supplier:  
Evolution® High-Column Workstations, Kewaunee®
Evolution® High-Column Workstations, Kewaunee®
Wallaby plastic laminate work surface, 36", 72×30"
Discontinued
Product Details & Documents
With a maximum height of 193 cm (76"), this workstation allows room for overhead storage, shelving, and lights.
  • The station features a front radius edge work surface plus two supporting columns with bases and glides
Starter workbenches include two columns, a modesty panel, front apron, worksurface brackets, and all hardware. Add-on workstation includes all items noted above except one supporting column and base; this workstation shares the column and base from an existing starter workbench.
Work surfaces are available in black epoxy resin, ESD almond static-dissipative laminate with grounding kit, and wallaby plastic laminate.
Depth: 30"
 : Starter workbenches and add-on workstation do not include a work surface. Work surfaces are available separately. A starter workbench is required before purchasing an add-on workstation. Standard color is light neutral with a black accent strip; or other color options, contact your Avantor sales representative.
 : Support brackets, an apron, and a modesty panel are included.
Specifications
  • Description:
    Wallaby plastic laminate work surface
  • Height:
    36"
  • W×D:
    72×30"