Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BASE/NEU EXTRACTABLES MIX4X1ML
Catalog # ULSVM-110
Supplier:
Ultra Scientific
SDS
Expand image
BASE/NEU EXTRACTABLES MIX4X1ML
Catalog # ULSVM-110
Supplier:
Ultra Scientific
Supplier Number:
SVM-110
SDS
Related Information
Documentation
SDS
ghs_SVM-110
Documentation