Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MIN SPIRITS SOL (25% WEA)4X1ML
Catalog # ULRGO-702
Supplier:
Ultra Scientific
SDS
Expand image
MIN SPIRITS SOL (25% WEA)4X1ML
Catalog # ULRGO-702
Supplier:
Ultra Scientific
Supplier Number:
RGO-702
SDS
Related Information
Documentation
SDS
ghs_RGO-702
Documentation