$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
KEROSENE (75% WEAT) 4X1ML
ULRGO-624
:
Ultra Scientific
KEROSENE (75% WEAT) 4X1ML
ULRGO-624
:
Ultra Scientific
:
RGO-624
ghs_RGO-624