$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
DIESEL FUEL #2 SOL (UNWE 4X1ML
ULRGO-611
:
Ultra Scientific
DIESEL FUEL #2 SOL (UNWE 4X1ML
ULRGO-611
:
Ultra Scientific
:
RGO-611
ghs_RGO-611