$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
UNLEADED GAS SLN (50% WE 4X1ML
ULRGO-603
:
Ultra Scientific
UNLEADED GAS SLN (50% WE 4X1ML
ULRGO-603
:
Ultra Scientific
:
RGO-603
ghs_RGO-603