$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
METHYLATED SURROGATE 4X1ML
ULPPS-262
:
Ultra Scientific
METHYLATED SURROGATE 4X1ML
ULPPS-262
:
Ultra Scientific
:
PPS-262
ghs_PPS-262