Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1,4-DIOXANE/100UG/ML METH4X1ML
Catalog # ULNV-150
Supplier:
Ultra Scientific
SDS
Expand image
1,4-DIOXANE/100UG/ML METH4X1ML
Catalog # ULNV-150
Supplier:
Ultra Scientific
Supplier Number:
NV-150
SDS
Related Information
Documentation
SDS
ghs_NV-150
Documentation