Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ACIDS SURR STD MIX 4X1ML
Catalog # ULISM-297
Supplier:
Ultra Scientific
SDS
Expand image
ACIDS SURR STD MIX 4X1ML
Catalog # ULISM-297
Supplier:
Ultra Scientific
Supplier Number:
ISM-297
SDS
Related Information
Documentation
SDS
ghs_ISM-297
Documentation