$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
SILVEX;2,4,5-TP 4X1ML
ULHB-110
:
Ultra Scientific
SILVEX;2,4,5-TP 4X1ML
ULHB-110
:
Ultra Scientific
:
HB-110
ghs_HB-110