$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CHLORINATED HYDROCARBONS 4X1ML
ULCHM-612
:
Ultra Scientific
CHLORINATED HYDROCARBONS 4X1ML
ULCHM-612
:
Ultra Scientific
:
CHM-612
ghs_CHM-612