$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CL6ETHANE 100PPM/MEOH 4X1ML
ULCH-180
:
Ultra Scientific
CL6ETHANE 100PPM/MEOH 4X1ML
ULCH-180
:
Ultra Scientific
:
CH-180
ghs_CH-180