$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CONCENTRATION CALIBRATIO 4X1ML
ULCB-681MN
:
Ultra Scientific
CONCENTRATION CALIBRATIO 4X1ML
ULCB-681MN
:
Ultra Scientific
:
CB-681MN
ghs_CB-681MN