$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
M-XYLENE-100UG/ML-METH 4X1ML
Catalog # ULAM-310
Supplier:
Ultra Scientific
SDS
Expand image
M-XYLENE-100UG/ML-METH 4X1ML
Catalog # ULAM-310
Supplier:
Ultra Scientific
Supplier Number:
AM-310
SDS
Related Information
Documentation
SDS
ghs_AM-310
Documentation