Wright-Giemsa stain
Glass bottle, 500 ml
Supplier: Ricca Chemical
: R9380000-500C
Jump to:
Product Details & Documents
Wright-Giemsa Stain Mixture
Synonyms: Wright-Giemsa Solution
- MDL Number:MFCD00132593
- Boiling Pt:~ 64.5 °C
- Density:0.8484 g/cm³
- Flash Pt:~ 12 °C (MeOH)
- Storage Temperature:Ambient
- UN:1993
- ADR:3,II
Specifications
Specification Test Results
- Absorbance (traceable to NIST SRM 930):Conforms to control
- Appearance:Dark blue, Methanol odor
Specifications
- Pack type:Glass bottle
- Size:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



