Formaldehyde 10% in aqueous solution stabilised histological fixative, buffered (pH 7.0)
Poly bottle, 2.5 gal.
Supplier: Ricca Chemical
Supplier #: R3190000-10F
Formaldehyde 10% in aqueous solution stabilised histological fixative, buffered (pH 7.0)
Catalog # RC319025
Supplier: Ricca Chemical
CAS Number: 50-00-0
Jump to:
Product Details & Documents
Contains neutral phosphate buffer.
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Storage Temperature:Ambient
- EINECS:200-001-8
- UN:3334
- ADR:9,
Documents
Specifications
Specification Test Results
- Appearance:Clear, colorless, Formaldehyde odor
- pH at 25 °C (traceable to NIST SRM 186 & 191):7.05 ± 0.05 pH at 25.0 °C
Specifications
- Pack type:Poly bottle
- Size:2.5 gal.
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



