Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
fixative reagent

fixative reagent

Bouin's Fixative Solution, Poly Bottle, 32 oz.
Catalog # RC112032
Supplier:   Ricca Chemical
Supplier #:  R1120000-1A
fixative reagent
Catalog # RC112032
Supplier:  Ricca Chemical
CAS Number:  
 fixative reagent
fixative reagent

Bouin's Fixative Solution, Poly Bottle, 32 oz.

CAS Number:  
Price
$144.61
Log in for availability
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Picric acid-formalin-acetic acid mixture.

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Poly Bottle
  • Description:
    Bouin's Fixative Solution
  • Size:
    32 oz.

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.