$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
AUTOCLAVE MODEL2540EP120V/60HZ
MP-023210411
:
Heidolph NA, LLC
AUTOCLAVE MODEL2540EP120V/60HZ
MP-023210411
:
Heidolph NA, LLC
:
023210410