Thymic Factor Trifluoroacetate
25 mg
Catalog # H-4865.0025BA
Supplier: Bachem Americas
Supplier #: 4009914.0025
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.
For exerting its anti-inflammatory and immunomodulatory activity, the peptide hormone thymulin, pEAKSQGGSN, requires the presence of Zn(II). Thymulin showed promising results in experimental models of lung diseases as well as potential as an analgesic peptide.
Specifications
Specifications
- Size:25 mg
- Storage Conditions:–20 ± 5 °C
- Cat. No.:H-4865.0025BA
- Molecular Weight:858.86
- Protein Synonyms:STF|Thymulin|Serum Thymic Factor
- Protein/Peptide Name:Thymic Factor Trifluoroacetate
- MDL No.:MFCD18782269
- Sequence:Pyr-Ala-Lys-Ser-Gln-Gly-Gly-Ser-Asn-OH
- Molecular Formula:C₃₃H₅₄N₁₂O₁₅
- CAS No.:63958-90-7
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



