Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Comparators for CHEMets® Test Kits

Comparators for CHEMets® Test Kits

Filming Amines Round Comparator, 0 - 1 ppm, 0, 0.05, 0.10, 0.15, 0.25, 0.50, 0.75, 1.0 ppm
Catalog # CXC-1001
Supplier:   CHEMetrics
Supplier #:  C-1001
Comparators for CHEMets® Test Kits
Catalog # CXC-1001
Supplier:  CHEMetrics
CAS Number:  
Comparators for CHEMets® Test Kits
Comparators for CHEMets® Test Kits

Filming Amines Round Comparator, 0 - 1 ppm, 0, 0.05, 0.10, 0.15, 0.25, 0.50, 0.75, 1.0 ppm

CAS Number:  
Price
$27.69
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

 : Store in the dark at room temperature.

Specifications

Specifications

  • Description:
    Filming Amines Round Comparator
  • Increments:
    0, 0.05, 0.10, 0.15, 0.25, 0.50, 0.75, 1.0 ppm
  • Range:
    0 - 1 ppm

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.