Orders LinkContactUsLinkComponent
Fmoc-D-Lys(Alloc)-OH
25 g
  B-2900.0025BA
 :  4027977.0025
Fmoc-D-Lys(Alloc)-OH
  B-2900.0025BA
 :  214750-75-1
Fmoc-D-Lys(Alloc)-OH
25 g
 :  214750-75-1
$758.01
Each (25g)
Sequence: Fmoc-D-Lys(Aloc)-OH
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
 :  Nepsilon-Allyloxycarbonyl-Nα-Fmoc-D-lysine, Fmoc-D-Lys(Aloc)-OH
  •  :214750-75-1
  •  :C₂₅H₂₈N₂O₆
  •  :MFCD00798639
  •  :452.507 g/mol
  •  :Freezer
  • Size:
    25 g