Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Fmoc-D-Lys(Alloc)-OH

Fmoc-D-Lys(Alloc)-OH

5 g
Catalog # B-2900.0005BA
Supplier:   Bachem Americas
Supplier #:  4027977.0005
Fmoc-D-Lys(Alloc)-OH
Catalog # B-2900.0005BA
Supplier:  Bachem Americas
CAS Number:  214750-75-1
Fmoc-D-Lys(Alloc)-OH

5 g

CAS Number:  214750-75-1
Price
$181.21
Log in for availability
Each (5g)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Sequence: Fmoc-D-Lys(Aloc)-OH

For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Synonyms:  Nepsilon-Allyloxycarbonyl-Nα-Fmoc-D-lysine, Fmoc-D-Lys(Aloc)-OH
  • CAS Number:214750-75-1
  • Formula:C₂₅H₂₈N₂O₆
  • MDL Number:MFCD00798639
  • Molecular Weight:452.507 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Size:
    5 g

Specifications