Fmoc-Dap(Alloc)-OH
5 g
Catalog # B-2845.0005BA
Supplier: Bachem Americas
Supplier #: 4026639.0005
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Sequence: Fmoc-Dap(Aloc)-OH
Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.
Bachem also offers Fmoc-Dap(Dnp)-SASRIN resin (D-2590) for the SPPS of FRET substrates.
Synonyms: Fmoc-Dap(Aloc)-OH, (S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid
- CAS Number:188970-92-5
- Formula:C₂₂H₂₂N₂O₆
- MDL Number:MFCD00273468
- Molecular Weight:410.426 g/mol
- Storage Temperature:Refrigerator
Specifications
Specifications
- Size:5 g



