Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Boc-Lys(Fmoc)-OH

25 g
Catalog # A-1610.0025BA
Supplier:   Bachem Americas
 :  4008005.0025
Boc-Lys(Fmoc)-OH
Catalog # A-1610.0025BA
Supplier:  Bachem Americas
CAS Number:  84624-27-1
Boc-Lys(Fmoc)-OH

25 g

CAS Number:  84624-27-1
Price
$1,302.17
Log in for availability
Each (25g)
 
Product Details & Documents

Sequence: Boc-Lys(Fmoc)-OH

For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Synonyms:  N-epsilon-FMOC-N-alpha-BOC-L-Lysine
  • CAS Number:84624-27-1
  • Formula:C₂₆H₃₂N₂O₆
  • MDL Number:MFCD00062032
  • Molecular Weight:468.55 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Size:
    25 g

Specifications