Orders LinkContactUsLinkComponent
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv
Safety Crimp Cap, 20 mm, 10 mm Hole, Aluminum, Silicone White/Aluminum Foil, 125 mil, Silver
Catalog # 97054-974
Headspace Caps, MicroSolv
Catalog # 97054-974
Supplier:  
Headspace Caps, MicroSolv
Headspace Caps, MicroSolv
Safety Crimp Cap, 20 mm, 10 mm Hole, Aluminum, Silicone White/Aluminum Foil, 125 mil, Silver
Discontinued
Product Details & Documents
MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.
The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.
Specifications
  • Septum:
    Silicone White/Aluminum Foil, 125 mil
  • Color:
    Silver
  • Description:
    Safety Crimp Cap
  • Material:
    Aluminum
  • Size:
    20 mm, 10 mm Hole